Assay Details
Gene Symbol: COX6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097473.2 | NP_001090942.2 | 2 - 3 | 213 | 134 | |
XM_013996570.1 | XP_013852024.1 | 191 | 134 | |||
GenBank mRNA | AK344443.1 | - | 2 - 3 | 178 | 134 | |
AK392221.1 | - | 2 - 3 | 183 | 134 | ||
DQ629154.1 | - | 1 - 2 | 143 | 134 | ||
HX218717.1 | - | 2 - 3 | 214 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
