Assay Details
Gene Symbol: COX6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097473.2 | NP_001090942.2 | 3 - 4 | 332 | 112 | |
XM_013996570.1 | XP_013852024.1 | 310 | 112 | |||
GenBank mRNA | AK344443.1 | - | 3 - 4 | 297 | 112 | |
AK392221.1 | - | 3 - 4 | 302 | 112 | ||
DQ629154.1 | - | 2 - 3 | 262 | 112 | ||
HX218717.1 | - | 3 - 4 | 333 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
