Assay Details
Gene Symbol: PCD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097491.1 | NP_001090960.1 | 3 - 4 | 609 | 55 | |
XM_021088429.1 | XP_020944088.1 | 734 | 55 | |||
GenBank mRNA | AB189910.1 | - | 3 - 4 | 609 | 55 | |
AK238838.1 | - | 3 - 4 | 655 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
