Assay Details
| Gene Symbol: CD1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097492.1 | NP_001090961.1 | 3 - 4 | 637 | 69 | |
| XM_005663233.3 | XP_005663290.1 | 1463 | 69 | |||
| GenBank mRNA | AB189912.1 | - | 3 - 4 | 637 | 69 | |
| AK352055.1 | - | 3 - 4 | 763 | 69 | ||
More Information
Set Membership: |
|

CD1E
Genomic Map