Assay Details
| Gene Symbol: CD72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097493.1 | NP_001090962.1 | 4 - 5 | 341 | 56 | |
| XM_005660185.3 | XP_005660242.1 | 1989 | 56 | |||
| XM_021096459.1 | XP_020952118.1 | 490 | 56 | |||
| XM_021097074.1 | XP_020952733.1 | 491 | 56 | |||
| GenBank mRNA | AK237013.1 | - | 4 - 5 | 471 | 56 | |
| AK348760.1 | - | 4 - 5 | 462 | 56 | ||
| DQ439546.1 | - | 4 - 5 | 341 | 56 | ||
| DQ439547.1 | - | 4 - 5 | 341 | 56 | ||
More Information
Set Membership: |
|

CD72
Genomic Map