Assay Details
Gene Symbol: PGRMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097521.2 | NP_001090990.2 | 2 - 3 | 589 | 87 | |
GenBank mRNA | AK236095.1 | - | 2 - 3 | 588 | 87 | |
AK400669.1 | - | 2 - 3 | 588 | 87 | ||
EF421714.1 | - | 3 - 4 | 390 | 87 | ||
EU242513.1 | - | 2 - 3 | 589 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
