Assay Details
Gene Symbol: EMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099940.1 | NP_001093410.1 | 1 - 2 | 77 | 60 | |
XM_021090848.1 | XP_020946507.1 | 300 | 60 | |||
XM_021090850.1 | XP_020946509.1 | 404 | 60 | |||
GenBank mRNA | AK391109.1 | - | 2 - 3 | 193 | 60 | |
EF577081.1 | - | 1 - 2 | 77 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
