Assay Details
| Gene Symbol: EIF4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100193.1 | NP_001093663.1 | 7 - 8 | 746 | 58 | |
| GenBank mRNA | DQ351286.1 | - | 7 - 8 | 746 | 58 | |
More Information
Set Membership: |
|

EIF4A3
Genomic Map