Assay Details
Gene Symbol: BPIFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101032.1 | NP_001094502.1 | 7 - 8 | 750 | 55 | |
XM_005672827.3 | XP_005672884.1 | 840 | 55 | |||
XM_021077173.1 | XP_020932832.1 | 841 | 55 | |||
GenBank mRNA | AK230859.1 | - | 7 - 8 | 733 | 55 | |
AK238078.1 | - | 7 - 8 | 742 | 55 | ||
AK238231.1 | - | 8 - 9 | 841 | 55 | ||
DQ859015.1 | - | 7 - 8 | 750 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
