Assay Details
Gene Symbol: NEU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101822.1 | NP_001095292.1 | 2 - 3 | 496 | 76 | |
GenBank mRNA | EU282367.1 | - | 2 - 3 | 362 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
