Assay Details
| Gene Symbol: DXO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101826.1 | NP_001095296.1 | 4 - 5 | 1010 | 57 | |
| XM_005665805.3 | XP_005665862.1 | 1183 | 57 | |||
| GenBank mRNA | AK351293.1 | - | 4 - 5 | 1172 | 57 | |
| EU282361.1 | - | 3 - 4 | 898 | 57 | ||
More Information
Set Membership: |
|

DXO
Genomic Map