Assay Details
Gene Symbol: NIFK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101827.1 | NP_001095297.1 | 6 - 7 | 753 | 92 | |
XM_021074506.1 | XP_020930165.1 | 813 | 92 | |||
GenBank mRNA | AK236623.1 | - | 6 - 7 | 776 | 92 | |
AK395871.1 | - | 6 - 7 | 775 | 92 | ||
AK399119.1 | - | 6 - 7 | 770 | 92 | ||
AY683049.1 | - | 6 - 7 | 753 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
