Assay Details
Gene Symbol: BCAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110175.1 | NP_001103645.1 | 2 - 3 | 153 | 62 | |
XM_021077178.1 | XP_020932837.1 | 489 | 62 | |||
XM_021077179.1 | XP_020932838.1 | 497 | 62 | |||
XM_021077180.1 | XP_020932839.1 | 497 | 62 | |||
XM_021077181.1 | XP_020932840.1 | 492 | 62 | |||
XM_021077182.1 | XP_020932841.1 | 499 | 62 | |||
XM_021077183.1 | XP_020932842.1 | 530 | 62 | |||
XM_021077184.1 | XP_020932843.1 | 484 | 62 | |||
XM_021077186.1 | XP_020932845.1 | 537 | 62 | |||
XM_021077187.1 | XP_020932846.1 | 541 | 62 | |||
XM_021077188.1 | XP_020932847.1 | 532 | 62 | |||
XM_021077189.1 | XP_020932848.1 | 473 | 62 | |||
XM_021077190.1 | XP_020932849.1 | 477 | 62 | |||
XM_021077191.1 | XP_020932850.1 | 441 | 62 | |||
XM_021077192.1 | XP_020932851.1 | 529 | 62 | |||
XM_021077193.1 | XP_020932852.1 | 528 | 62 | |||
XM_021077194.1 | XP_020932853.1 | 517 | 62 | |||
XM_021077195.1 | XP_020932854.1 | 524 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
