Assay Details
| Gene Symbol: CSTF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110430.1 | NP_001103900.1 | 6 - 7 | 1199 | 95 | |
| XM_005673029.3 | XP_005673086.1 | 1353 | 95 | |||
| XM_005673030.3 | XP_005673087.1 | 1473 | 95 | |||
| XM_021077196.1 | XP_020932855.1 | 1523 | 95 | |||
| GenBank mRNA | AK235656.1 | - | 6 - 7 | 1183 | 95 | |
| AK239052.1 | - | 6 - 7 | 1198 | 95 | ||
| AK396372.1 | - | 6 - 7 | 1213 | 95 | ||
More Information
Set Membership: |
|

CSTF1
Genomic Map