Assay Details
| Gene Symbol: CASS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110431.2 | NP_001103901.2 | 2 - 3 | 594 | 108 | |
| XM_013985477.2 | XP_013840931.2 | 1033 | 108 | |||
More Information
Set Membership: |
|

CASS4
Genomic Map