Assay Details
| Gene Symbol: RTF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110432.1 | NP_001103902.1 | 1 - 2 | 167 | 69 | |
| XM_005673032.3 | XP_005673089.1 | 154 | 69 | |||
| GenBank mRNA | AK233642.1 | - | 1 - 2 | 117 | 69 | |
| AK235189.1 | - | 1 - 2 | 140 | 69 | ||
| AK348007.1 | - | 1 - 2 | 93 | 69 | ||
| AK396192.1 | - | 1 - 2 | 139 | 69 | ||
| AK397035.1 | - | 1 - 2 | 143 | 69 | ||
| AK397163.1 | - | 1 - 2 | 142 | 69 | ||
| AK399170.1 | - | 1 - 2 | 142 | 69 | ||
More Information
Set Membership: |
|

RTF2
Genomic Map