Assay Details
| Gene Symbol: RTF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110432.1 | NP_001103902.1 | 7 - 8 | 743 | 101 | |
| XM_005673032.3 | XP_005673089.1 | 730 | 101 | |||
| GenBank mRNA | AK235189.1 | - | 7 - 8 | 716 | 101 | |
| AK348007.1 | - | 7 - 8 | 669 | 101 | ||
| AK397035.1 | - | 7 - 8 | 719 | 101 | ||
| AK397163.1 | - | 7 - 8 | 718 | 101 | ||
| AK399170.1 | - | 7 - 8 | 718 | 101 | ||
More Information
Set Membership: |
|

RTF2
Genomic Map