Assay Details
Gene Symbol: CIDEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112688.1 | NP_001106158.1 | 2 - 3 | 248 | 62 | |
GenBank mRNA | AK232542.1 | - | 2 - 3 | 295 | 62 | |
EF374061.1 | - | 2 - 3 | 248 | 62 | ||
EU143328.1 | - | 2 - 3 | 246 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
