Assay Details
Gene Symbol: CIDEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112688.1 | NP_001106158.1 | 3 - 4 | 395 | 62 | |
XM_021097868.1 | XP_020953527.1 | 676 | 62 | |||
GenBank mRNA | AK232542.1 | - | 3 - 4 | 442 | 62 | |
EF374061.1 | - | 3 - 4 | 395 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
