Assay Details
Gene Symbol: CIDEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112688.1 | NP_001106158.1 | 4 - 5 | 578 | 90 | |
XM_021097868.1 | XP_020953527.1 | 859 | 90 | |||
GenBank mRNA | AK232542.1 | - | 4 - 5 | 625 | 90 | |
EF374061.1 | - | 4 - 5 | 578 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
