Assay Details
Gene Symbol: PLA2G7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113013.2 | NP_001106484.1 | 3 - 4 | 439 | 73 | |
NM_001322806.1 | NP_001309735.1 | 3 - 4 | 391 | 73 | ||
NM_001322807.1 | NP_001309736.1 | 2 - 3 | 296 | 73 | ||
XM_005666084.3 | XP_005666141.1 | 378 | 73 | |||
XM_013977998.2 | XP_013833452.1 | 456 | 73 | |||
XM_013977999.2 | XP_013833453.1 | 333 | 73 | |||
XM_021098277.1 | XP_020953936.1 | 477 | 73 | |||
XM_021098278.1 | XP_020953937.1 | 274 | 73 | |||
XM_021098280.1 | XP_020953939.1 | 322 | 73 | |||
GenBank mRNA | AK235478.1 | - | 2 - 3 | 212 | 73 | |
AK393913.1 | - | 3 - 4 | 391 | 73 | ||
AK395076.1 | - | 2 - 3 | 237 | 73 | ||
AK396677.1 | - | 4 - 5 | 348 | 73 | ||
AY610106.1 | - | 3 - 4 | 426 | 73 | ||
AY972538.1 | - | 2 - 3 | 226 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
