Assay Details
| Gene Symbol: KIR2DL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113218.1 | NP_001106689.1 | 4 - 5 | 699 | 70 | |
| XM_021093201.1 | XP_020948860.1 | 1235 | 70 | |||
More Information
Set Membership: |
|

KIR2DL1
Genomic Map