Assay Details
| Gene Symbol: SLA-DRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113695.1 | NP_001107167.1 | 3 - 4 | 726 | 70 | |
| GenBank mRNA | AB016746.1 | - | 2 - 3 | 567 | 70 | |
| AB215120.1 | - | 3 - 4 | 654 | 70 | ||
| AB215124.1 | - | 3 - 4 | 654 | 70 | ||
| AF464034.1 | - | 3 - 4 | 654 | 70 | ||
| AF464046.2 | - | 3 - 4 | 647 | 70 | ||
| AK237255.1 | - | 4 - 5 | 634 | 70 | ||
| AK347078.1 | - | 3 - 4 | 696 | 70 | ||
| AK348716.1 | - | 3 - 4 | 711 | 70 | ||
| AK348721.1 | - | 3 - 4 | 717 | 70 | ||
| AK350429.1 | - | 3 - 4 | 717 | 70 | ||
| AK351376.1 | - | 3 - 4 | 721 | 70 | ||
| AK394000.1 | - | 3 - 4 | 719 | 70 | ||
| AK396034.1 | - | 3 - 4 | 718 | 70 | ||
| AK396070.1 | - | 3 - 4 | 722 | 70 | ||
| AK396087.1 | - | 3 - 4 | 720 | 70 | ||
| AK398292.1 | - | 3 - 4 | 724 | 70 | ||
| AK398847.1 | - | 3 - 4 | 743 | 70 | ||
| AK398993.1 | - | 3 - 4 | 722 | 70 | ||
| AK399081.1 | - | 3 - 4 | 719 | 70 | ||
| AY135576.1 | - | 2 - 3 | 364 | 70 | ||
| AY135579.1 | - | 3 - 4 | 726 | 70 | ||
| AY135580.1 | - | 3 - 4 | 614 | 70 | ||
| AY962313.1 | - | 3 - 4 | 656 | 70 | ||
| DQ272492.1 | - | 3 - 4 | 654 | 70 | ||
| DQ303220.1 | - | 3 - 4 | 70 | |||
| DQ883227.1 | - | 3 - 4 | 675 | 70 | ||
| EU431221.1 | - | 4 - 5 | 675 | 70 | ||
| EU432072.1 | - | 3 - 4 | 656 | 70 | ||
| EU432076.1 | - | 3 - 4 | 656 | 70 | ||
| EU432079.1 | - | 3 - 4 | 656 | 70 | ||
| EU722920.1 | - | 3 - 4 | 675 | 70 | ||
| KJ776456.1 | - | 3 - 4 | 654 | 70 | ||
| KU754598.1 | - | 3 - 4 | 675 | 70 | ||
| KU754600.1 | - | 3 - 4 | 675 | 70 | ||
| M55165.1 | - | 2 - 3 | 690 | 70 | ||
| Gene Symbol: LOC106504372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315758.1 | NP_001302687.1 | 3 - 4 | 654 | 70 | |
| GenBank mRNA | AF464033.2 | - | 3 - 4 | 654 | 70 | |
| AY135578.1 | - | 3 - 4 | 714 | 70 | ||
More Information
Set Membership: |
|

SLA-DRB1
Genomic Map