Assay Details
| Gene Symbol: GCNT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113699.1 | NP_001107171.1 | 2 - 3 | 1130 | 68 | |
| XM_005673033.3 | XP_005673090.1 | 1941 | 68 | |||
More Information
Set Membership: |
|

GCNT7
Genomic Map