Assay Details
| Gene Symbol: SLA-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113703.1 | NP_001107175.1 | 1 - 2 | 59 | 57 | |
| GenBank mRNA | AK231632.1 | - | 1 - 2 | 105 | 57 | |
| AK233184.1 | - | 1 - 2 | 77 | 57 | ||
| AK235295.1 | - | 1 - 2 | 77 | 57 | ||
| AK389292.1 | - | 1 - 2 | 111 | 57 | ||
| AK397717.1 | - | 1 - 2 | 86 | 57 | ||
| AY463542.1 | - | 1 - 2 | 59 | 57 | ||
| GU322909.1 | - | 1 - 2 | 59 | 57 | ||
More Information
Set Membership: |
|

SLA-8
Genomic Map