Assay Details
| Gene Symbol: CCNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114282.1 | NP_001107754.1 | 5 - 6 | 594 | 64 | |
| XM_021082593.1 | XP_020938252.1 | 731 | 64 | |||
| GenBank mRNA | AB332237.1 | - | 5 - 6 | 594 | 64 | |
More Information
Set Membership: |
|

CCNB2
Genomic Map