Assay Details
Gene Symbol: ADNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123071.1 | NP_001116543.1 | 3 - 4 | 846 | 67 | |
XM_013985467.2 | XP_013840921.1 | 2570 | 67 | |||
XM_021077204.1 | XP_020932863.1 | 1927 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
