Assay Details
Gene Symbol: C4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123089.2 | NP_001116561.1 | 17 - 18 | 2316 | 89 | |
GenBank mRNA | EU076594.1 | - | 17 - 18 | 2317 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
