Assay Details
Gene Symbol: C4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123089.2 | NP_001116561.1 | 29 - 30 | 3947 | 93 | |
GenBank mRNA | AY349423.1 | - | 1 - 2 | 224 | 93 | |
EU076594.1 | - | 29 - 30 | 3948 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
