Assay Details
Gene Symbol: C4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123089.2 | NP_001116561.1 | 31 - 32 | 4176 | 85 | |
GenBank mRNA | AK232400.1 | - | 2 - 3 | 196 | 85 | |
AY349423.1 | - | 3 - 4 | 453 | 85 | ||
EU076594.1 | - | 31 - 32 | 4177 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
