Assay Details
Gene Symbol: C4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123089.2 | NP_001116561.1 | 36 - 37 | 4730 | 59 | |
GenBank mRNA | AK232400.1 | - | 7 - 8 | 750 | 59 | |
AK350147.1 | - | 5 - 6 | 563 | 59 | ||
EU076594.1 | - | 36 - 37 | 4731 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
