Assay Details
Gene Symbol: C4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123089.2 | NP_001116561.1 | 9 - 10 | 1093 | 56 | |
GenBank mRNA | AK344119.1 | - | 9 - 10 | 1055 | 56 | |
EU076594.1 | - | 9 - 10 | 1094 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
