Assay Details
Gene Symbol: CTSZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123104.1 | NP_001116576.1 | 3 - 4 | 624 | 59 | |
GenBank mRNA | AK234297.1 | - | 3 - 4 | 585 | 59 | |
AK237527.1 | - | 3 - 4 | 550 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
