Assay Details
Gene Symbol: CTSZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123104.1 | NP_001116576.1 | 5 - 6 | 929 | 73 | |
GenBank mRNA | AK234297.1 | - | 5 - 6 | 890 | 73 | |
AK237527.1 | - | 5 - 6 | 855 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
