Assay Details
Gene Symbol: ITM2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123131.1 | NP_001116603.1 | 4 - 5 | 563 | 56 | |
GenBank mRNA | AK237361.1 | - | 4 - 5 | 529 | 56 | |
AK392188.1 | - | 4 - 5 | 775 | 56 | ||
AK398775.1 | - | 4 - 5 | 666 | 56 | ||
DQ917621.1 | - | 4 - 5 | 563 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
