Assay Details
| Gene Symbol: MC3R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123137.1 | NP_001116609.1 | 1 | 954 | 57 | |
| GenBank mRNA | JQ828976.1 | - | 1 | 857 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MC3R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123137.1 | NP_001116609.1 | 1 | 954 | 57 | |
| GenBank mRNA | JQ828976.1 | - | 1 | 857 | 57 | |
Set Membership: |
|