Assay Details
Gene Symbol: PMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123163.1 | NP_001116635.1 | 3 - 4 | 370 | 64 | |
GenBank mRNA | AK231871.1 | - | 3 - 4 | 380 | 64 | |
EU534185.1 | - | 3 - 4 | 370 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
