Assay Details
| Gene Symbol: PPP1R11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123166.1 | NP_001116638.1 | 1 - 2 | 71 | 73 | |
| GenBank mRNA | AK234087.1 | - | 1 - 2 | 161 | 73 | |
| AK237611.1 | - | 1 - 2 | 248 | 73 | ||
| AK392734.1 | - | 1 - 2 | 228 | 73 | ||
More Information
Set Membership: |
|

PPP1R11
Genomic Map