Assay Details
Gene Symbol: PMEPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123203.1 | NP_001116675.1 | 2 - 3 | 686 | 64 | |
XM_005673042.3 | XP_005673099.1 | 364 | 64 | |||
XM_013985486.2 | XP_013840940.1 | 313 | 64 | |||
GenBank mRNA | AK398064.1 | - | 2 - 3 | 412 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
