Assay Details
| Gene Symbol: ZBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123216.1 | NP_001116688.1 | 6 - 7 | 1014 | 63 | |
| GenBank mRNA | AK350043.1 | - | 6 - 7 | 1034 | 63 | |
| FJ455510.1 | - | 6 - 7 | 941 | 63 | ||
More Information
Set Membership: |
|

ZBP1
Genomic Map