Assay Details
| Gene Symbol: AGER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123218.1 | NP_001116690.1 | 10 - 11 | 1190 | 59 | |
| XM_013977655.2 | XP_013833109.1 | 1205 | 59 | |||
| XM_013977656.2 | XP_013833110.1 | 993 | 59 | |||
| GenBank mRNA | EU282357.1 | - | 10 - 11 | 1080 | 59 | |
| GU164706.1 | - | 8 - 9 | 1558 | 59 | ||
| GU164707.1 | - | 7 - 8 | 1512 | 59 | ||
| GU164708.1 | - | 8 - 9 | 1488 | 59 | ||
| GU164709.1 | - | 8 - 9 | 1402 | 59 | ||
| GU164711.1 | - | 9 - 10 | 1308 | 59 | ||
| GU164712.1 | - | 8 - 9 | 1197 | 59 | ||
| GU164713.1 | - | 9 - 10 | 947 | 59 | ||
| GU164714.1 | - | 3 - 4 | 886 | 59 | ||
More Information
Set Membership: |
|

AGER
Genomic Map