Assay Details
| Gene Symbol: CREB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123220.1 | NP_001116692.1 | 4 - 5 | 438 | 60 | |
| XM_005660189.3 | XP_005660246.2 | 930 | 60 | |||
| XM_005660190.3 | XP_005660247.2 | 583 | 60 | |||
| XM_013993639.2 | XP_013849093.2 | 589 | 60 | |||
| GenBank mRNA | AK235744.1 | - | 4 - 5 | 539 | 60 | |
| AK389376.1 | - | 5 - 6 | 551 | 60 | ||
| AK390381.1 | - | 5 - 6 | 748 | 60 | ||
| AK393011.1 | - | 4 - 5 | 567 | 60 | ||
| AK400220.1 | - | 4 - 5 | 600 | 60 | ||
| EU588585.1 | - | 4 - 5 | 495 | 60 | ||
More Information
Set Membership: |
|

CREB3
Genomic Map