Assay Details
| Gene Symbol: CRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213844.2 | NP_999009.1 | 1 - 2 | 153 | 110 | |
| GenBank mRNA | AB005545.1 | - | 1 - 2 | 174 | 110 | |
| AY714055.1 | - | 1 - 2 | 153 | 110 | ||
| EU812512.1 | - | 1 - 2 | 63 | 110 | ||
More Information
Set Membership: |
|

CRP
Genomic Map