Assay Details
| Gene Symbol: MAN1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213885.1 | NP_999050.1 | 9 - 10 | 1874 | 74 | |
| XM_021082358.1 | XP_020938017.1 | 2111 | 74 | |||
| GenBank mRNA | Y12503.1 | - | 1874 | 74 | ||
More Information
Set Membership: |
|

MAN1A1
Genomic Map