Assay Details
| Gene Symbol: VCAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213891.1 | NP_999056.1 | 4 - 5 | 1024 | 69 | |
| GenBank mRNA | AK235268.1 | - | 4 - 5 | 1034 | 69 | |
| L43124.1 | - | 4 - 5 | 1016 | 69 | ||
| U08351.1 | - | 4 - 5 | 1024 | 69 | ||
More Information
Set Membership: |
|

VCAM1
Genomic Map