Assay Details
Gene Symbol: H3-3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213930.1 | NP_999095.1 | 3 - 4 | 344 | 86 | |
GenBank mRNA | AF469469.1 | - | 3 - 4 | 344 | 86 | |
AK233751.1 | - | 3 - 4 | 398 | 86 | ||
AK233980.1 | - | 3 - 4 | 406 | 86 | ||
AK240156.1 | - | 806 | 86 | |||
AK240322.1 | - | 3 - 4 | 405 | 86 | ||
AK348103.1 | - | 3 - 4 | 397 | 86 | ||
AK392212.1 | - | 3 - 4 | 406 | 86 | ||
AK392304.1 | - | 3 - 4 | 401 | 86 | ||
AK393004.1 | - | 3 - 4 | 406 | 86 | ||
AK394431.1 | - | 3 - 4 | 398 | 86 | ||
AK396090.1 | - | 4 - 5 | 517 | 86 | ||
AK398832.1 | - | 3 - 4 | 406 | 86 | ||
AK399089.1 | - | 3 - 4 | 396 | 86 | ||
AY610300.1 | - | 3 - 4 | 351 | 86 |
Gene Symbol: LOC100627410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021100000.1 | XP_020955659.1 | 529 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
