Assay Details
| Gene Symbol: SMAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213965.1 | NP_999130.1 | 3 - 4 | 1023 | 62 | |
| XM_021100448.1 | XP_020956107.1 | 1630 | 62 | |||
| XM_021100449.1 | XP_020956108.1 | 981 | 62 | |||
| XM_021100451.1 | XP_020956110.1 | 1555 | 62 | |||
| XM_021100452.1 | XP_020956111.1 | 1483 | 62 | |||
| XM_021100453.1 | XP_020956112.1 | 4394 | 62 | |||
| XM_021100454.1 | XP_020956113.1 | 1191 | 62 | |||
| XM_021100455.1 | XP_020956114.1 | 855 | 62 | |||
| GenBank mRNA | AK235916.1 | - | 3 - 4 | 1094 | 62 | |
| AK397691.1 | - | 4 - 5 | 1156 | 62 | ||
| AK400271.1 | - | 3 - 4 | 1162 | 62 | ||
| AY245888.1 | - | 3 - 4 | 1023 | 62 | ||
More Information
Set Membership: |
|

SMAD1
Genomic Map