Assay Details
| Gene Symbol: HSP90AA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213973.2 | NP_999138.1 | 2111 | 98 | ||
| GenBank mRNA | AK234540.1 | - | 528 | 98 | ||
| AK237363.1 | - | 477 | 98 | |||
| AK240418.1 | - | 965 | 98 | |||
| AK392141.1 | - | 2272 | 98 | |||
| AK398738.1 | - | 439 | 98 | |||
| U94395.1 | - | 2111 | 98 | |||
More Information
Set Membership: |
|

HSP90AA1
Genomic Map