Assay Details
| Gene Symbol: SPAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214011.1 | NP_999176.1 | 3 - 4 | 1346 | 87 | |
| GenBank mRNA | AJ310675.2 | - | 3 - 4 | 1346 | 87 | |
More Information
Set Membership: |
|

SPAM1
Genomic Map