Assay Details
| Gene Symbol: ZP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214045.1 | NP_999210.1 | 10 - 11 | 1426 | 81 | |
| GenBank mRNA | L11000.1 | - | 10 - 11 | 1426 | 81 | |
More Information
Set Membership: |
|

ZP4
Genomic Map